Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616375 | ||||||||||||||||||||
Literature Links: |
MIR4738 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR4738 - microRNA 4738 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UNK - unkempt family zinc finger | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080419.2 | 412 | Intron | NP_001073888.2 | |||
XM_006722145.3 | 412 | UTR 5 | XP_006722208.1 | |||
XM_006722148.2 | 412 | Intron | XP_006722211.1 | |||
XM_011525400.1 | 412 | Intron | XP_011523702.1 | |||
XM_011525401.2 | 412 | Intron | XP_011523703.1 | |||
XM_017025247.1 | 412 | UTR 5 | XP_016880736.1 | |||
XM_017025248.1 | 412 | Intron | XP_016880737.1 | |||
XM_017025249.1 | 412 | UTR 5 | XP_016880738.1 | |||
XM_017025250.1 | 412 | Intron | XP_016880739.1 |