Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176261 MIM: 602917 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCNE1 - potassium voltage-gated channel subfamily E regulatory subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RCAN1 - regulator of calcineurin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001285389.2 | 2103 | UTR 3 | NP_001272318.1 | |||
NM_001285391.2 | 2103 | UTR 3 | NP_001272320.2 | |||
NM_001285392.2 | 2103 | UTR 3 | NP_001272321.1 | |||
NM_001285393.2 | 2103 | UTR 3 | NP_001272322.1 | |||
NM_004414.6 | 2103 | UTR 3 | NP_004405.3 | |||
NM_203417.2 | 2103 | UTR 3 | NP_981962.1 | |||
NM_203418.2 | 2103 | UTR 3 | NP_981963.1 | |||
XM_005260929.1 | 2103 | Intron | XP_005260986.1 | |||
XM_005260930.1 | 2103 | Intron | XP_005260987.1 | |||
XM_017028282.1 | 2103 | Intron | XP_016883771.1 | |||
XM_017028283.1 | 2103 | UTR 3 | XP_016883772.1 |