Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616909 MIM: 603869 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC68 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC68 - coiled-coil domain containing 68 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143829.1 | 2528 | UTR 3 | NP_001137301.1 | |||
NM_025214.2 | 2528 | UTR 3 | NP_079490.1 | |||
XM_006722552.3 | 2528 | UTR 3 | XP_006722615.1 | |||
XM_011526198.2 | 2528 | UTR 3 | XP_011524500.1 | |||
XM_011526199.2 | 2528 | UTR 3 | XP_011524501.1 | |||
XM_011526200.2 | 2528 | UTR 3 | XP_011524502.1 | |||
XM_011526201.2 | 2528 | UTR 3 | XP_011524503.1 | |||
XM_011526202.1 | 2528 | UTR 3 | XP_011524504.1 | |||
XM_011526203.2 | 2528 | UTR 3 | XP_011524505.1 | |||
XM_011526204.2 | 2528 | Intron | XP_011524506.1 | |||
XM_017026011.1 | 2528 | UTR 3 | XP_016881500.1 |
RAB27B - RAB27B, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |