Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAGGGAAGGGTTGGGCAGAGAGA[G/T]GAGGGGCAGCATCAGTGCAGCTGGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 135620 MIM: 609124 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITGA5 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ITGA5 - integrin subunit alpha 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002205.4 | 4139 | UTR 3 | NP_002196.4 |
LOC102724050 - uncharacterized LOC102724050 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF385A - zinc finger protein 385A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |