Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605248 | ||||||||||||||||||||
Literature Links: |
MCOLN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MCOLN1 - mucolipin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF358 - zinc finger protein 358 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018083.4 | 1206 | Missense Mutation | AAG,AGG | K,R 382 | NP_060553.4 | |
XM_005272460.3 | 1206 | Missense Mutation | AAG,AGG | K,R 382 | XP_005272517.1 | |
XM_011527695.1 | 1206 | Missense Mutation | AAG,AGG | K,R 382 | XP_011525997.1 | |
XM_017026296.1 | 1206 | Missense Mutation | AAG,AGG | K,R 401 | XP_016881785.1 |