Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601011 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CACNA1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CACNA1A - calcium voltage-gated channel subunit alpha1 A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000068.3 | 6596 | Silent Mutation | ACC,ACG | T,T 2120 | NP_000059.3 | |
NM_001127221.1 | 6596 | Silent Mutation | ACC,ACG | T,T 2115 | NP_001120693.1 | |
NM_001127222.1 | 6596 | Silent Mutation | ACC,ACG | T,T 2114 | NP_001120694.1 | |
NM_001174080.1 | 6596 | Silent Mutation | ACC,ACG | T,T 2117 | NP_001167551.1 | |
NM_023035.2 | 6596 | Silent Mutation | ACC,ACG | T,T 2120 | NP_075461.2 |