Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603340 | ||||||||||||||||||||
Literature Links: |
ASB14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASB14 - ankyrin repeat and SOCS box containing 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DNAH12 - dynein axonemal heavy chain 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291661.1 | 11588 | Missense Mutation | AGA,GGA | R,G 3808 | NP_001278590.1 | |
NM_198564.3 | 11588 | Intron | NP_940966.2 | |||
XM_011533466.1 | 11588 | Missense Mutation | AGA,GGA | R,G 3804 | XP_011531768.1 | |
XM_011533467.1 | 11588 | Missense Mutation | AGA,GGA | R,G 3793 | XP_011531769.1 | |
XM_011533468.1 | 11588 | Missense Mutation | AGA,GGA | R,G 3783 | XP_011531770.1 | |
XM_011533469.1 | 11588 | Missense Mutation | AGA,GGA | R,G 3757 | XP_011531771.1 | |
XM_011533471.2 | 11588 | Intron | XP_011531773.1 | |||
XM_011533474.1 | 11588 | Intron | XP_011531776.1 | |||
XM_017005860.1 | 11588 | Intron | XP_016861349.1 | |||
XM_017005861.1 | 11588 | Missense Mutation | AGA,GGA | R,G 3304 | XP_016861350.1 | |
XM_017005862.1 | 11588 | Missense Mutation | AGA,GGA | R,G 2668 | XP_016861351.1 | |
XM_017005863.1 | 11588 | Intron | XP_016861352.1 |