Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606781 | ||||||||||||||||||||
Literature Links: |
TKT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TKT - transketolase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001064.3 | 1935 | Missense Mutation | CTG,GTG | L,V 601 | NP_001055.1 | |
NM_001135055.2 | 1935 | Missense Mutation | CTG,GTG | L,V 601 | NP_001128527.1 | |
NM_001258028.1 | 1935 | Missense Mutation | CTG,GTG | L,V 609 | NP_001244957.1 | |
XM_011534054.1 | 1935 | Missense Mutation | CTG,GTG | L,V 609 | XP_011532356.1 | |
XM_011534055.1 | 1935 | Missense Mutation | CTG,GTG | L,V 435 | XP_011532357.1 |