Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607010 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCP1A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DCP1A - decapping mRNA 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290204.1 | 4140 | UTR 3 | NP_001277133.1 | |||
NM_001290205.1 | 4140 | UTR 3 | NP_001277134.1 | |||
NM_001290206.1 | 4140 | UTR 3 | NP_001277135.1 | |||
NM_001290207.1 | 4140 | UTR 3 | NP_001277136.1 | |||
NM_018403.6 | 4140 | UTR 3 | NP_060873.4 |
Set Membership: |
HapMap |