Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602242 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP2S1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AP2S1 - adaptor related protein complex 2 sigma 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301076.1 | Intron | NP_001288005.1 | ||||
NM_001301078.1 | Intron | NP_001288007.1 | ||||
NM_001301081.1 | Intron | NP_001288010.1 | ||||
NM_004069.4 | Intron | NP_004060.2 | ||||
NM_021575.3 | Intron | NP_067586.1 | ||||
XM_011526423.2 | Intron | XP_011524725.1 | ||||
XM_011526424.2 | Intron | XP_011524726.1 |
Set Membership: |
HapMap |