Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MEIS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MEIS3 - Meis homeobox 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009813.2 | 2110 | Intron | NP_001009813.1 | |||
NM_001301059.1 | 2110 | Intron | NP_001287988.1 | |||
NM_020160.2 | 2110 | Intron | NP_064545.1 | |||
XM_011527136.2 | 2110 | UTR 3 | XP_011525438.1 | |||
XM_011527138.2 | 2110 | UTR 3 | XP_011525440.1 | |||
XM_011527139.2 | 2110 | Intron | XP_011525441.1 | |||
XM_011527140.1 | 2110 | UTR 3 | XP_011525442.1 | |||
XM_017027011.1 | 2110 | UTR 3 | XP_016882500.1 |
Set Membership: |
HapMap |