Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
52 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602144 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRDT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRDT - bromodomain testis associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242805.2 | 39 | UTR 5 | NP_001229734.2 | |||
NM_001242806.2 | 39 | UTR 5 | NP_001229735.2 | |||
NM_001242807.2 | 39 | UTR 5 | NP_001229736.2 | |||
NM_001242808.2 | 39 | UTR 5 | NP_001229737.2 | |||
NM_001242810.2 | 39 | UTR 5 | NP_001229739.2 | |||
NM_001726.4 | 39 | Intron | NP_001717.3 | |||
NM_207189.3 | 39 | UTR 5 | NP_997072.2 | |||
XM_006710853.3 | 39 | UTR 5 | XP_006710916.1 | |||
XM_006710854.3 | 39 | UTR 5 | XP_006710917.1 | |||
XM_006710855.3 | 39 | UTR 5 | XP_006710918.1 | |||
XM_006710856.3 | 39 | UTR 5 | XP_006710919.1 | |||
XM_006710857.3 | 39 | UTR 5 | XP_006710920.1 | |||
XM_011542032.2 | 39 | UTR 5 | XP_011540334.1 | |||
XM_011542033.2 | 39 | UTR 5 | XP_011540335.1 | |||
XM_011542034.2 | 39 | UTR 5 | XP_011540336.1 | |||
XM_011542035.2 | 39 | UTR 5 | XP_011540337.1 | |||
XM_011542036.2 | 39 | UTR 5 | XP_011540338.1 |