Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602296 MIM: 614729 MIM: 600592 MIM: 612983 MIM: 612150 MIM: 612984 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP4M1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AP4M1 - adaptor related protein complex 4 mu 1 subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
COPS6 - COP9 signalosome subunit 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006833.4 | 2671 | Intron | NP_006824.2 |
MCM7 - minichromosome maintenance complex component 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278595.1 | 2671 | Silent Mutation | GTC,GTG | V,V 499 | NP_001265524.1 | |
NM_005916.4 | 2671 | Silent Mutation | GTC,GTG | V,V 675 | NP_005907.3 | |
NM_182776.2 | 2671 | Silent Mutation | GTC,GTG | V,V 499 | NP_877577.1 | |
XM_005250348.3 | 2671 | Silent Mutation | GTC,GTG | V,V 568 | XP_005250405.1 | |
XM_017012217.1 | 2671 | Silent Mutation | GTC,GTG | V,V 568 | XP_016867706.1 |
MIR106B - microRNA 106b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR25 - microRNA 25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR93 - microRNA 93 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |