Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607754 | ||||||||||||||||||||
Literature Links: |
MKRN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MKRN1 - makorin ring finger protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145125.1 | 1126 | Intron | NP_001138597.1 | |||
NM_001291663.1 | 1126 | Intron | NP_001278592.1 | |||
NM_013446.3 | 1126 | Missense Mutation | AAG,AGG | K,R 371 | NP_038474.2 | |
XM_011515996.1 | 1126 | Missense Mutation | AAG,AGG | K,R 328 | XP_011514298.1 | |
XM_011515997.2 | 1126 | Missense Mutation | AAG,AGG | K,R 307 | XP_011514299.1 | |
XM_011515998.1 | 1126 | Intron | XP_011514300.1 |