Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACTCAGACCACATTAGCAATGT[G/T]TATAAAAGGGAGCAATTTAATCATT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604764 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C8orf76 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C8orf76 - chromosome 8 open reading frame 76 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZHX1 - zinc fingers and homeoboxes 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017926.2 | 4905 | UTR 3 | NP_001017926.1 | |||
NM_007222.4 | 4905 | UTR 3 | NP_009153.3 |
ZHX1-C8orf76 - ZHX1-C8orf76 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204180.1 | 4905 | Intron | NP_001191109.1 |
Set Membership: |
HapMap |