Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 604743 | ||||||||||||||||||||
Literature Links: |
DDAH1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DDAH1 - dimethylarginine dimethylaminohydrolase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134445.1 | 3839 | UTR 3 | NP_001127917.1 | |||
NM_012137.3 | 3839 | UTR 3 | NP_036269.1 | |||
XM_005270707.2 | 3839 | UTR 3 | XP_005270764.1 | |||
XM_005270710.2 | 3839 | UTR 3 | XP_005270767.1 | |||
XM_011541158.1 | 3839 | UTR 3 | XP_011539460.1 | |||
XM_017000889.1 | 3839 | UTR 3 | XP_016856378.1 | |||
XM_017000890.1 | 3839 | UTR 3 | XP_016856379.1 |