Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 609696 | |||||||||||||||||||||||||||||
Literature Links: |
ARID4B PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese)
|
|||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ARID4B - AT-rich interaction domain 4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206794.1 | 5508 | UTR 3 | NP_001193723.1 | |||
NM_016374.5 | 5508 | UTR 3 | NP_057458.4 | |||
NM_031371.3 | 5508 | UTR 3 | NP_112739.2 | |||
XM_006711781.2 | 5508 | UTR 3 | XP_006711844.1 | |||
XM_011544212.2 | 5508 | UTR 3 | XP_011542514.1 | |||
XM_017001468.1 | 5508 | UTR 3 | XP_016856957.1 | |||
XM_017001469.1 | 5508 | UTR 3 | XP_016856958.1 | |||
XM_017001470.1 | 5508 | UTR 3 | XP_016856959.1 | |||
XM_017001471.1 | 5508 | Intron | XP_016856960.1 | |||
XM_017001472.1 | 5508 | UTR 3 | XP_016856961.1 |
RBM34 - RNA binding motif protein 34 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |