Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTCCAGCTCTCGCGAGACTTTCA[A/G]GGGTCGGAGCGCGGGGGCCGGCCGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 616574 | ||||||||||||||||||||
Literature Links: |
LOC105378614 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC105378614 - uncharacterized LOC105378614 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MINOS1 - mitochondrial inner membrane organizing system 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032363.3 | 15 | UTR 5 | NP_001027535.1 | |||
NM_001204082.1 | 15 | UTR 5 | NP_001191011.1 | |||
NM_001204083.1 | 15 | UTR 5 | NP_001191012.1 |
MINOS1-NBL1 - MINOS1-NBL1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204088.1 | 15 | UTR 5 | NP_001191017.1 | |||
NM_001204089.1 | 15 | UTR 5 | NP_001191018.1 |