Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605405 | ||||||||||||||||||||
Literature Links: |
USP6NL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
USP6NL - USP6 N-terminal like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080491.3 | 7515 | UTR 3 | NP_001073960.1 | |||
NM_014688.3 | 7515 | UTR 3 | NP_055503.1 | |||
XM_006717542.3 | 7515 | UTR 3 | XP_006717605.1 | |||
XM_011519762.2 | 7515 | UTR 3 | XP_011518064.1 | |||
XM_011519763.2 | 7515 | UTR 3 | XP_011518065.1 | |||
XM_017016971.1 | 7515 | UTR 3 | XP_016872460.1 | |||
XM_017016972.1 | 7515 | UTR 3 | XP_016872461.1 | |||
XM_017016973.1 | 7515 | UTR 3 | XP_016872462.1 |