Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
42 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SH3BP5L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SH3BP5L - SH3 binding domain protein 5 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322462.1 | 1795 | UTR 3 | NP_001309391.1 | |||
NM_001322463.1 | 1795 | UTR 3 | NP_001309392.1 | |||
NM_001322464.1 | 1795 | UTR 3 | NP_001309393.1 | |||
NM_030645.1 | 1795 | UTR 3 | NP_085148.1 | |||
XM_017002404.1 | 1795 | UTR 3 | XP_016857893.1 | |||
XM_017002405.1 | 1795 | UTR 3 | XP_016857894.1 | |||
XM_017002406.1 | 1795 | UTR 3 | XP_016857895.1 | |||
XM_017002407.1 | 1795 | UTR 3 | XP_016857896.1 |
Set Membership: |
Validated |