Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613365 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLCO6A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLCO6A1 - solute carrier organic anion transporter family member 6A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289002.1 | Intron | NP_001275931.1 | ||||
NM_001289004.1 | Intron | NP_001275933.1 | ||||
NM_001308014.1 | Intron | NP_001294943.1 | ||||
NM_173488.4 | Intron | NP_775759.3 | ||||
XM_005271874.3 | Intron | XP_005271931.1 | ||||
XM_011543147.2 | Intron | XP_011541449.1 | ||||
XM_011543148.2 | Intron | XP_011541450.1 | ||||
XM_011543150.1 | Intron | XP_011541452.1 | ||||
XM_011543153.2 | Intron | XP_011541455.1 | ||||
XM_011543154.2 | Intron | XP_011541456.1 |
Set Membership: |
HapMap |