Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGATGATGTTGCTGGCGGCTCT[A/G]AGATCTGCAAGGGGCAGAGAAAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610537 MIM: 604346 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPP7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DPP7 - dipeptidyl peptidase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013379.2 | 1242 | Silent Mutation | CTC,CTT | L,L 404 | NP_037511.2 | |
XM_005266075.3 | 1242 | Silent Mutation | CTC,CTT | L,L 477 | XP_005266132.1 | |
XM_006717083.3 | 1242 | Nonsense Mutation | CAG,TAG | Q,* 455 | XP_006717146.1 | |
XM_011518599.2 | 1242 | Silent Mutation | CTC,CTT | L,L 426 | XP_011516901.1 | |
XM_011518600.1 | 1242 | Silent Mutation | CTC,CTT | L,L 396 | XP_011516902.1 | |
XM_017014651.1 | 1242 | Nonsense Mutation | CAG,TAG | Q,* 404 | XP_016870140.1 | |
XM_017014652.1 | 1242 | Nonsense Mutation | CAG,TAG | Q,* 382 | XP_016870141.1 |
LOC101930307 - uncharacterized LOC101930307 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAN1B1 - mannosidase alpha class 1B member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |