Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612111 | ||||||||||||||||||||
Literature Links: |
TNFAIP8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TNFAIP8 - TNF alpha induced protein 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077654.2 | Intron | NP_001071122.1 | ||||
NM_001286813.1 | Intron | NP_001273742.1 | ||||
NM_001286814.1 | Intron | NP_001273743.1 | ||||
NM_001286815.1 | Intron | NP_001273744.1 | ||||
NM_001286817.1 | Intron | NP_001273746.1 | ||||
NM_014350.3 | Intron | NP_055165.2 | ||||
XM_017009327.1 | Intron | XP_016864816.1 | ||||
XM_017009328.1 | Intron | XP_016864817.1 |