Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118495 MIM: 601441 MIM: 162096 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRM4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CHRM4 - cholinergic receptor muscarinic 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DGKZ - diacylglycerol kinase zeta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MDK - midkine (neurite growth-promoting factor 2) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012333.2 | 430 | Silent Mutation | CTA,CTG | L,L 14 | NP_001012333.1 | |
NM_001012334.2 | 430 | Silent Mutation | CTA,CTG | L,L 14 | NP_001012334.1 | |
NM_001270550.1 | 430 | Silent Mutation | CTA,CTG | L,L 14 | NP_001257479.1 | |
NM_001270551.1 | 430 | Silent Mutation | CTA,CTG | L,L 14 | NP_001257480.1 | |
NM_001270552.1 | 430 | Silent Mutation | CTA,CTG | L,L 14 | NP_001257481.1 | |
NM_002391.4 | 430 | Silent Mutation | CTA,CTG | L,L 14 | NP_002382.1 | |
XM_011520116.2 | 430 | Silent Mutation | CTA,CTG | L,L 14 | XP_011518418.1 | |
XM_017017764.1 | 430 | Silent Mutation | CTA,CTG | L,L 14 | XP_016873253.1 |
MIR4688 - microRNA 4688 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |