Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGAAGCTGAGCTCCCTAGTCTA[C/G]GAGAGGGTGGCACTTGCTGTAGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611211 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGEF17 - Rho guanine nucleotide exchange factor 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RELT - RELT tumor necrosis factor receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032871.3 | Intron | NP_116260.2 | ||||
NM_152222.1 | Intron | NP_689408.1 | ||||
XM_005274356.2 | Intron | XP_005274413.1 | ||||
XM_006718714.2 | Intron | XP_006718777.1 | ||||
XM_011545305.1 | Intron | XP_011543607.1 |