Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGACTGAATAAAACCTGATCAAT[A/G]TAATTTCTCTGTCAGTTCATATTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179513 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB6A PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
RAB6A - RAB6A, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243718.1 | Intron | NP_001230647.1 | ||||
NM_001243719.1 | Intron | NP_001230648.1 | ||||
NM_002869.4 | Intron | NP_002860.2 | ||||
NM_198896.1 | Intron | NP_942599.1 |