Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604503 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
JMJD1C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
JMJD1C - jumonji domain containing 1C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282948.1 | 7833 | Missense Mutation | GAC,GAG | D,E 2353 | NP_001269877.1 | |
NM_001318153.1 | 7833 | Missense Mutation | GAC,GAG | D,E 2247 | NP_001305082.1 | |
NM_001318154.1 | 7833 | Missense Mutation | GAC,GAG | D,E 2353 | NP_001305083.1 | |
NM_001322252.1 | 7833 | Missense Mutation | GAC,GAG | D,E 2497 | NP_001309181.1 | |
NM_001322254.1 | 7833 | Missense Mutation | GAC,GAG | D,E 2316 | NP_001309183.1 | |
NM_001322258.1 | 7833 | Missense Mutation | GAC,GAG | D,E 2316 | NP_001309187.1 | |
NM_032776.2 | 7833 | Missense Mutation | GAC,GAG | D,E 2535 | NP_116165.1 | |
XM_011539503.1 | 7833 | Missense Mutation | GAC,GAG | D,E 2312 | XP_011537805.1 | |
XM_011539508.2 | 7833 | Intron | XP_011537810.1 | |||
XM_017015897.1 | 7833 | Missense Mutation | GAC,GAG | D,E 2353 | XP_016871386.1 | |
XM_017015898.1 | 7833 | Missense Mutation | GAC,GAG | D,E 2353 | XP_016871387.1 | |
XM_017015899.1 | 7833 | Missense Mutation | GAC,GAG | D,E 2247 | XP_016871388.1 | |
XM_017015900.1 | 7833 | Missense Mutation | GAC,GAG | D,E 2247 | XP_016871389.1 | |
XM_017015901.1 | 7833 | Missense Mutation | GAC,GAG | D,E 2247 | XP_016871390.1 | |
XM_017015902.1 | 7833 | Missense Mutation | GAC,GAG | D,E 2247 | XP_016871391.1 | |
XM_017015903.1 | 7833 | Missense Mutation | GAC,GAG | D,E 2247 | XP_016871392.1 |
Set Membership: |
HapMap Validated |