Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612189 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PBLD PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PBLD - phenazine biosynthesis like protein domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033083.1 | Intron | NP_001028255.1 | ||||
NM_022129.3 | Intron | NP_071412.2 | ||||
XM_005270028.3 | Intron | XP_005270085.1 | ||||
XM_011540060.2 | Intron | XP_011538362.1 | ||||
XM_017016513.1 | Intron | XP_016872002.1 | ||||
XM_017016514.1 | Intron | XP_016872003.1 |