Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCGCCGCCGATGCGGCCGTCGGG[A/G]AGCGCCTGGAGGTGGAAGCCGATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164980 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGF4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FGF4 - fibroblast growth factor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002007.2 | 616 | Silent Mutation | CTC,CTT | L,L 99 | NP_001998.1 | |
XM_005273847.1 | 616 | Silent Mutation | CTC,CTT | L,L 99 | XP_005273904.1 |