Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603817 MIM: 607153 | ||||||||||||||||||||
Literature Links: |
ASAP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASAP2 - ArfGAP with SH3 domain, ankyrin repeat and PH domain 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ITGB1BP1 - integrin subunit beta 1 binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319066.1 | 1444 | UTR 3 | NP_001305995.1 | |||
NM_001319067.1 | 1444 | UTR 3 | NP_001305996.1 | |||
NM_001319068.1 | 1444 | UTR 3 | NP_001305997.1 | |||
NM_001319069.1 | 1444 | UTR 3 | NP_001305998.1 | |||
NM_001319070.1 | 1444 | UTR 3 | NP_001305999.1 | |||
NM_004763.4 | 1444 | UTR 3 | NP_004754.1 | |||
NM_022334.4 | 1444 | UTR 3 | NP_071729.1 | |||
XM_005246183.4 | 1444 | UTR 3 | XP_005246240.1 | |||
XM_005246184.4 | 1444 | UTR 3 | XP_005246241.1 | |||
XM_005246185.4 | 1444 | UTR 3 | XP_005246242.1 | |||
XM_005246189.4 | 1444 | UTR 3 | XP_005246246.1 | |||
XM_006711903.3 | 1444 | UTR 3 | XP_006711966.1 | |||
XM_011510416.2 | 1444 | UTR 3 | XP_011508718.1 | |||
XM_017005267.1 | 1444 | UTR 3 | XP_016860756.1 | |||
XM_017005268.1 | 1444 | UTR 3 | XP_016860757.1 | |||
XM_017005269.1 | 1444 | UTR 3 | XP_016860758.1 | |||
XM_017005270.1 | 1444 | UTR 3 | XP_016860759.1 |