Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603817 MIM: 607153 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASAP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ASAP2 - ArfGAP with SH3 domain, ankyrin repeat and PH domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135191.1 | 1618 | Intron | NP_001128663.1 | |||
NM_003887.2 | 1618 | Intron | NP_003878.1 | |||
XM_006711898.1 | 1618 | Intron | XP_006711961.1 | |||
XM_006711901.3 | 1618 | Intron | XP_006711964.1 | |||
XM_006711902.1 | 1618 | Intron | XP_006711965.1 | |||
XM_011510403.2 | 1618 | Intron | XP_011508705.1 | |||
XM_011510404.2 | 1618 | Intron | XP_011508706.1 | |||
XM_011510405.2 | 1618 | Intron | XP_011508707.1 | |||
XM_011510406.2 | 1618 | Intron | XP_011508708.1 | |||
XM_011510407.2 | 1618 | Intron | XP_011508709.1 | |||
XM_011510408.2 | 1618 | Intron | XP_011508710.1 | |||
XM_011510409.1 | 1618 | Intron | XP_011508711.1 |
ITGB1BP1 - integrin subunit beta 1 binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319066.1 | 1618 | UTR 3 | NP_001305995.1 | |||
NM_001319067.1 | 1618 | UTR 3 | NP_001305996.1 | |||
NM_001319068.1 | 1618 | UTR 3 | NP_001305997.1 | |||
NM_001319069.1 | 1618 | UTR 3 | NP_001305998.1 | |||
NM_001319070.1 | 1618 | UTR 3 | NP_001305999.1 | |||
NM_004763.4 | 1618 | UTR 3 | NP_004754.1 | |||
NM_022334.4 | 1618 | UTR 3 | NP_071729.1 | |||
XM_005246183.4 | 1618 | Intron | XP_005246240.1 | |||
XM_005246184.4 | 1618 | Intron | XP_005246241.1 | |||
XM_005246185.4 | 1618 | Intron | XP_005246242.1 | |||
XM_005246189.4 | 1618 | Intron | XP_005246246.1 | |||
XM_006711903.3 | 1618 | Intron | XP_006711966.1 | |||
XM_011510416.2 | 1618 | Intron | XP_011508718.1 | |||
XM_017005267.1 | 1618 | Intron | XP_016860756.1 | |||
XM_017005268.1 | 1618 | Intron | XP_016860757.1 | |||
XM_017005269.1 | 1618 | Intron | XP_016860758.1 | |||
XM_017005270.1 | 1618 | Intron | XP_016860759.1 |