Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603549 MIM: 176740 MIM: 617019 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDS2 - CDP-diacylglycerol synthase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCNA - proliferating cell nuclear antigen | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002592.2 | 734 | Silent Mutation | GAC,GAT | D,D 165 | NP_002583.1 | |
NM_182649.1 | 734 | Intron | NP_872590.1 |
PCNA-AS1 - PCNA antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM230 - transmembrane protein 230 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |