Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601439 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCC9 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
ABCC9 - ATP binding cassette subfamily C member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005691.3 | 6497 | Intron | NP_005682.2 | |||
NM_020297.3 | 6497 | UTR 3 | NP_064693.2 | |||
XM_005253284.3 | 6497 | UTR 3 | XP_005253341.1 | |||
XM_005253286.3 | 6497 | UTR 3 | XP_005253343.1 | |||
XM_005253287.4 | 6497 | UTR 3 | XP_005253344.1 | |||
XM_005253288.3 | 6497 | UTR 3 | XP_005253345.1 | |||
XM_005253289.3 | 6497 | UTR 3 | XP_005253346.1 | |||
XM_005253290.3 | 6497 | UTR 3 | XP_005253347.1 | |||
XM_006719025.3 | 6497 | UTR 3 | XP_006719088.1 | |||
XM_011520545.2 | 6497 | UTR 3 | XP_011518847.1 |
Set Membership: |
HapMap Validated |