Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAATGTGTGCCTCTACAGTAATGT[C/G]GGAAATAAATACCATTCTTGAAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615337 MIM: 604649 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
B3GNTL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
B3GNTL1 - UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009905.2 | 2014 | UTR 3 | NP_001009905.1 | |||
NM_001320742.1 | 2014 | UTR 3 | NP_001307671.1 | |||
NM_001320743.1 | 2014 | UTR 3 | NP_001307672.1 | |||
XM_006722272.3 | 2014 | Intron | XP_006722335.1 | |||
XM_006722274.2 | 2014 | Intron | XP_006722337.2 | |||
XM_011523532.2 | 2014 | Intron | XP_011521834.1 | |||
XM_011523535.1 | 2014 | Intron | XP_011521837.1 | |||
XM_017024205.1 | 2014 | Intron | XP_016879694.1 | |||
XM_017024206.1 | 2014 | Intron | XP_016879695.1 | |||
XM_017024207.1 | 2014 | Intron | XP_016879696.1 |
LOC105371944 - uncharacterized LOC105371944 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011523611.2 | 2014 | Intron | XP_011521913.1 |
TBCD - tubulin folding cofactor D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005993.4 | 2014 | UTR 3 | NP_005984.3 | |||
XM_005256396.4 | 2014 | Intron | XP_005256453.1 | |||
XM_005256399.4 | 2014 | UTR 3 | XP_005256456.1 | |||
XM_005256404.4 | 2014 | UTR 3 | XP_005256461.1 | |||
XM_006722290.2 | 2014 | UTR 3 | XP_006722353.1 | |||
XM_006722291.3 | 2014 | UTR 3 | XP_006722354.1 | |||
XM_006722292.2 | 2014 | UTR 3 | XP_006722355.1 | |||
XM_011523589.1 | 2014 | UTR 3 | XP_011521891.1 | |||
XM_011523590.1 | 2014 | UTR 3 | XP_011521892.1 | |||
XM_011523591.1 | 2014 | UTR 3 | XP_011521893.1 | |||
XM_011523593.1 | 2014 | UTR 3 | XP_011521895.1 | |||
XM_011523594.1 | 2014 | UTR 3 | XP_011521896.1 | |||
XM_011523595.2 | 2014 | UTR 3 | XP_011521897.1 | |||
XM_011523597.2 | 2014 | UTR 3 | XP_011521899.1 | |||
XM_011523598.1 | 2014 | UTR 3 | XP_011521900.1 | |||
XM_011523599.2 | 2014 | UTR 3 | XP_011521901.1 | |||
XM_011523600.2 | 2014 | UTR 3 | XP_011521902.1 | |||
XM_017024987.1 | 2014 | Intron | XP_016880476.1 | |||
XM_017024988.1 | 2014 | Intron | XP_016880477.1 | |||
XM_017024989.1 | 2014 | UTR 3 | XP_016880478.1 | |||
XM_017024990.1 | 2014 | UTR 3 | XP_016880479.1 |
Set Membership: |
HapMap |