Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 182392 | ||||||||||||||||||||
Literature Links: |
SCN7A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SCN7A - sodium voltage-gated channel alpha subunit 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002976.3 | 7326 | UTR 3 | NP_002967.2 | |||
XM_006712680.2 | 7326 | UTR 3 | XP_006712743.1 | |||
XM_006712681.3 | 7326 | UTR 3 | XP_006712744.1 | |||
XM_006712682.3 | 7326 | UTR 3 | XP_006712745.1 | |||
XM_011511615.2 | 7326 | UTR 3 | XP_011509917.1 | |||
XM_017004667.1 | 7326 | UTR 3 | XP_016860156.1 |