Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615295 MIM: 602559 | ||||||||||||||||||||
Literature Links: |
USP34 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
USP34 - ubiquitin specific peptidase 34 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
XPO1 - exportin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003400.3 | 4612 | UTR 3 | NP_003391.1 | |||
XM_005264544.1 | 4612 | UTR 3 | XP_005264601.1 | |||
XM_005264546.1 | 4612 | UTR 3 | XP_005264603.1 | |||
XM_006712094.2 | 4612 | UTR 3 | XP_006712157.1 | |||
XM_011533097.1 | 4612 | UTR 3 | XP_011531399.1 | |||
XM_011533098.1 | 4612 | UTR 3 | XP_011531400.1 | |||
XM_011533099.2 | 4612 | UTR 3 | XP_011531401.1 |