Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611433 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAGOHB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAGOHB - mago homolog B, exon junction complex core component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STYK1 - serine/threonine/tyrosine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018423.2 | Intron | NP_060893.2 | ||||
XM_005253417.2 | Intron | XP_005253474.1 | ||||
XM_011520736.1 | Intron | XP_011519038.1 | ||||
XM_011520737.2 | Intron | XP_011519039.1 | ||||
XM_011520738.2 | Intron | XP_011519040.1 | ||||
XM_017019579.1 | Intron | XP_016875068.1 |
Set Membership: |
HapMap |