Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612088 MIM: 606783 | ||||||||||||||||||||
Literature Links: |
CLEC12A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLEC12A - C-type lectin domain family 12 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001207010.1 | 1273 | Intron | NP_001193939.1 | |||
NM_001300730.1 | 1273 | Intron | NP_001287659.1 | |||
NM_138337.5 | 1273 | Intron | NP_612210.4 | |||
NM_201623.3 | 1273 | Intron | NP_963917.2 | |||
XM_005253324.2 | 1273 | Intron | XP_005253381.1 | |||
XM_006719033.3 | 1273 | Intron | XP_006719096.1 | |||
XM_006719035.3 | 1273 | Intron | XP_006719098.1 | |||
XM_006719036.3 | 1273 | Intron | XP_006719099.1 | |||
XM_011520570.1 | 1273 | Intron | XP_011518872.1 | |||
XM_011520571.2 | 1273 | Intron | XP_011518873.2 | |||
XM_011520573.2 | 1273 | Intron | XP_011518875.1 |
CLEC1B - C-type lectin domain family 1 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099431.1 | 1273 | UTR 3 | NP_001092901.1 | |||
NM_016509.3 | 1273 | UTR 3 | NP_057593.3 | |||
XM_005253382.4 | 1273 | UTR 3 | XP_005253439.1 | |||
XM_011520685.2 | 1273 | UTR 3 | XP_011518987.1 | |||
XM_011520686.2 | 1273 | Intron | XP_011518988.1 | |||
XM_017019395.1 | 1273 | UTR 3 | XP_016874884.1 | |||
XM_017019396.1 | 1273 | Intron | XP_016874885.1 |