Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600537 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PYROXD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PYROXD1 - pyridine nucleotide-disulphide oxidoreductase domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024854.3 | 1902 | Intron | NP_079130.2 | |||
XM_006719153.3 | 1902 | Intron | XP_006719216.1 | |||
XM_017019976.1 | 1902 | Intron | XP_016875465.1 |
RECQL - RecQ like helicase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002907.3 | 1902 | Missense Mutation | CAT,GAT | H,D 495 | NP_002898.2 | |
NM_032941.2 | 1902 | Missense Mutation | CAT,GAT | H,D 495 | NP_116559.1 | |
XM_005253461.3 | 1902 | Missense Mutation | CAT,GAT | H,D 495 | XP_005253518.1 | |
XM_005253462.4 | 1902 | Missense Mutation | CAT,GAT | H,D 495 | XP_005253519.1 | |
XM_005253463.3 | 1902 | Missense Mutation | CAT,GAT | H,D 495 | XP_005253520.1 | |
XM_005253464.3 | 1902 | Missense Mutation | CAT,GAT | H,D 495 | XP_005253521.1 |