Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602243 MIM: 614177 MIM: 601189 MIM: 602644 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD151 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CD151 - CD151 molecule (Raph blood group) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CRACR2B - calcium release activated channel regulator 2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLR2L - RNA polymerase II subunit L | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021128.4 | 120 | Silent Mutation | NP_066951.1 |
TSPAN4 - tetraspanin 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |