Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600702 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCN8A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SCN8A - sodium voltage-gated channel alpha subunit 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177984.2 | Intron | NP_001171455.1 | ||||
NM_014191.3 | Intron | NP_055006.1 | ||||
XM_006719556.3 | Intron | XP_006719619.1 | ||||
XM_011538650.2 | Intron | XP_011536952.1 | ||||
XM_011538651.2 | Intron | XP_011536953.1 | ||||
XM_017019794.1 | Intron | XP_016875283.1 | ||||
XM_017019795.1 | Intron | XP_016875284.1 | ||||
XM_017019796.1 | Intron | XP_016875285.1 |
Set Membership: |
HapMap |