Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608508 MIM: 603236 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYBA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CYBA - cytochrome b-245 alpha chain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MVD - mevalonate diphosphate decarboxylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002461.2 | 1446 | UTR 3 | NP_002452.1 | |||
XM_011523086.2 | 1446 | UTR 3 | XP_011521388.1 | |||
XM_011523087.2 | 1446 | UTR 3 | XP_011521389.1 | |||
XM_011523088.2 | 1446 | UTR 3 | XP_011521390.1 | |||
XM_011523089.2 | 1446 | UTR 3 | XP_011521391.1 |
Set Membership: |
HapMap |