Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
INTS6L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
INTS6L - integrator complex subunit 6 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182540.4 | Intron | NP_872346.3 | ||||
XM_005262394.2 | Intron | XP_005262451.1 | ||||
XM_006724740.2 | Intron | XP_006724803.1 | ||||
XM_006724741.3 | Intron | XP_006724804.1 | ||||
XM_011531311.2 | Intron | XP_011529613.1 | ||||
XM_017029342.1 | Intron | XP_016884831.1 | ||||
XM_017029343.1 | Intron | XP_016884832.1 | ||||
XM_017029344.1 | Intron | XP_016884833.1 | ||||
XM_017029345.1 | Intron | XP_016884834.1 | ||||
XM_017029346.1 | Intron | XP_016884835.1 | ||||
XM_017029347.1 | Intron | XP_016884836.1 | ||||
XM_017029348.1 | Intron | XP_016884837.1 | ||||
XM_017029349.1 | Intron | XP_016884838.1 |