Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCAGGATCGAAGCAGTGTGGTT[A/G]TTTTTCGGGTAATTCTCCATCTGCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601444 MIM: 155740 MIM: 601617 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BLOC1S1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BLOC1S1 - biogenesis of lysosomal organelles complex 1 subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
BLOC1S1-RDH5 - BLOC1S1-RDH5 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CD63 - CD63 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001257389.1 | 528 | Silent Mutation | AAC,AAT | N,N 129 | NP_001244318.1 | |
NM_001257390.1 | 528 | Silent Mutation | AAC,AAT | N,N 129 | NP_001244319.1 | |
NM_001257391.1 | 528 | Silent Mutation | AAC,AAT | N,N 129 | NP_001244320.1 | |
NM_001257392.1 | 528 | Silent Mutation | AAC,AAT | N,N 106 | NP_001244321.1 | |
NM_001257400.1 | 528 | Silent Mutation | AAC,AAT | N,N 47 | NP_001244329.1 | |
NM_001257401.1 | 528 | Silent Mutation | AAC,AAT | N,N 47 | NP_001244330.1 | |
NM_001267698.1 | 528 | Silent Mutation | AAC,AAT | N,N 129 | NP_001254627.1 | |
NM_001780.5 | 528 | Silent Mutation | AAC,AAT | N,N 129 | NP_001771.1 |
RDH5 - retinol dehydrogenase 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |