Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 154550 | ||||||||||||||||||||
Literature Links: |
FAM219B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM219B - family with sequence similarity 219 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321920.1 | 1519 | UTR 3 | NP_001308849.1 | |||
NM_001321921.1 | 1519 | Intron | NP_001308850.1 | |||
NM_001321922.1 | 1519 | UTR 3 | NP_001308851.1 | |||
NM_001321923.1 | 1519 | UTR 3 | NP_001308852.1 | |||
NM_020447.4 | 1519 | UTR 3 | NP_065180.1 | |||
XM_017022433.1 | 1519 | Intron | XP_016877922.1 |
MPI - mannose phosphate isomerase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |