Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
69 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf189 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
C1orf189 - chromosome 1 open reading frame 189 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010979.2 | 1289 | Intron | NP_001010979.1 |
C1orf43 - chromosome 1 open reading frame 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098616.2 | 1289 | UTR 3 | NP_001092086.1 | |||
NM_001297717.1 | 1289 | UTR 3 | NP_001284646.1 | |||
NM_001297718.1 | 1289 | UTR 3 | NP_001284647.1 | |||
NM_001297720.1 | 1289 | UTR 3 | NP_001284649.1 | |||
NM_001297721.1 | 1289 | Intron | NP_001284650.1 | |||
NM_001297723.1 | 1289 | Intron | NP_001284652.1 | |||
NM_015449.3 | 1289 | UTR 3 | NP_056264.1 | |||
NM_138740.3 | 1289 | UTR 3 | NP_620077.1 |