Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611409 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OCA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
OCA2 - OCA2 melanosomal transmembrane protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000275.2 | Intron | NP_000266.2 | ||||
NM_001300984.1 | Intron | NP_001287913.1 | ||||
XM_011521640.2 | Intron | XP_011519942.1 | ||||
XM_017022255.1 | Intron | XP_016877744.1 | ||||
XM_017022256.1 | Intron | XP_016877745.1 | ||||
XM_017022257.1 | Intron | XP_016877746.1 | ||||
XM_017022258.1 | Intron | XP_016877747.1 | ||||
XM_017022259.1 | Intron | XP_016877748.1 | ||||
XM_017022260.1 | Intron | XP_016877749.1 | ||||
XM_017022261.1 | Intron | XP_016877750.1 | ||||
XM_017022262.1 | Intron | XP_016877751.1 | ||||
XM_017022263.1 | Intron | XP_016877752.1 | ||||
XM_017022264.1 | Intron | XP_016877753.1 | ||||
XM_017022265.1 | Intron | XP_016877754.1 |
Set Membership: |
HapMap |