Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTAACAACTTGCTAAAAGAATGC[A/C]AATCTCACTTCTTCAGAGTAATAGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611740 MIM: 605791 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCO2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BCO2 - beta-carotene oxygenase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037290.2 | Intron | NP_001032367.2 | ||||
NM_001256397.1 | Intron | NP_001243326.1 | ||||
NM_001256398.1 | Intron | NP_001243327.1 | ||||
NM_001256400.1 | Intron | NP_001243329.1 | ||||
NM_031938.5 | Intron | NP_114144.4 |
TEX12 - testis expressed 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |