Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607240 MIM: 614093 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KMT5A PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KMT5A - lysine methyltransferase 5A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RILPL2 - Rab interacting lysosomal protein like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145058.2 | 1280 | UTR 3 | NP_659495.1 | |||
XM_011538012.2 | 1280 | Intron | XP_011536314.1 | |||
XM_017018959.1 | 1280 | Intron | XP_016874448.1 |