Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602504 | ||||||||||||||||||||
Literature Links: |
SHOX2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SHOX2 - short stature homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163678.1 | 1020 | UTR 3 | NP_001157150.1 | |||
NM_003030.4 | 1020 | UTR 3 | NP_003021.3 | |||
NM_006884.3 | 1020 | UTR 3 | NP_006875.2 | |||
XM_006713727.3 | 1020 | UTR 3 | XP_006713790.1 | |||
XM_006713728.3 | 1020 | UTR 3 | XP_006713791.1 | |||
XM_017007053.1 | 1020 | UTR 3 | XP_016862542.1 | |||
XM_017007054.1 | 1020 | UTR 3 | XP_016862543.1 | |||
XM_017007055.1 | 1020 | UTR 3 | XP_016862544.1 |